Folic acid ≥97%
Catalog #: 89151-408
Supplier: Enzo Life Sciences
Price
$75.40
Log in for availability
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:10 g
Specifications
Specification Test Results
- Folic acid ≥97%:
Specification Test Results
About this item
Synonyms: Vitamin M,(S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid,Pteroyl-L-glutamic acid,PteGlu,Vitamin B9
:Folic acid
:Length
:Width
:Height
:Diameter
:Thickness
:97
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 441.41 g/mol
Formula:C₁₉H₁₉N₇O₆
MW:441.41 g/mol
MDL Number:MFCD00079305
CAS Number:59-30-3
Customers Who Bought This Also Bought