Anti-ERBB2 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
810-227-L001
89155-690EA 2375.07 CAD
89155-690
Anti-ERBB2 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit, Isotype: IgG

  • For Immunohistochemistry

Recognizes human C-erbB-2.

Type: Primary
Antigen: ERBB2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR