Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

15-Deoxy-Δ12,14-Prostaglandin J2, Enzo Life Sciences

Supplier: Enzo Life Sciences

BML-PG050-0001 BML-PG050-0005
Prostaglandin D2 derived nuclear peroxisome proliferator-activated receptor γ (PPARγ) ligand. Induces apoptosis in a number of cell types including human lung cancer cells, synoviocytes and macrophages. Promotes differentiation of fibroblasts to adipocytes. Antiinflammatory activity. Inhibits angiogenesis. Prevents TNFα induced leptin increase in mice. TRPA1 agonist. Serious doubts as to the biological relevance of this prostaglandin have been raised. Endogenous levels seem to be too low to have any effect as a PPARγ ligand.

  • PPARγ ligand

Neat oil. Vials may appear empty because the oil tends to coat the inside of the vial.

Soluble in 100% ethanol (25mg/ml) or DMSO (25mg/ml).


Formula: C₂₀H₂₈O₃
MW: 316,441 g/mol
Storage Temperature: Freezer
CAS Number: 87893-55-8


 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
1 mg BML-PG050-0001 89161-528 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

5 mg BML-PG050-0005 89161-530 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping