Anti-Proteasome 11S Beta Subunit Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW8240-0025 PW8240-0100
89162-088EA 362.31 CAD
89162-088 89162-090
Anti-Proteasome 11S Beta Subunit Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse and rat proteasome activator 11S (beta subunit).

Type: Primary
Antigen: PROTEASOME 11S BETA SUBUNIT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR