Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TPH Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
TZ1050-0050
Tryptophan hydroxylase is regarded as the rate-limiting enzyme in serotonin (5-HT; 5-hydroxytryptamine) biosynthesis. TPH immunoreactivity is localised exclusively to serotonergic neurons in the central and peripheral nervous systems.

For Immunohistochemistry, Western Blot

Recognizes primate, rabbit, rat and cat tryptophan hydroxylase.

Type: Primary
Antigen: TPH
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Rat




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
50 µL TZ1050-0050 89165-072 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol TPH
Antigen name Tryptophan Hydroxylase
Reactivity Rat
Clonality Monoclonal
Antibody type Primary
Host Mouse
Isotype IgG3
Western blot Yes
ImmunoChemistry Yes
Antigen synonyms MGC119994, tryptophan 5-hydroxylase 1, TPRH, tryptophan hydroxylase (tryptophan 5-monooxygenase), TRPH, indoleacetic acid-5-hydroxylase, L-tryptophan hydroxylase, tryptophan 5-monooxygenase 1
Form Liquid. Ascites containing 0.1% sodium azide.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping