Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SUMO-2 (human), (recombinant) (agarose immobilized), Enzo Life Sciences

Supplier: Enzo Life Sciences
UW0100-0500
Human recombinant SUMO-2 coupled to agarose at 0.5mg protein per ml settled resin.

For isolation and enrichment of SUMO-2 interacting proteins. SUMO-2 agarose may have considerable utility for the capture/pull down of SUMO-2 interacting partners, including processing/signalling pathway proteins and SUMO-2 modification substrates, from cell lysates and extracts by affinity binding. The immobilized protein retains the exposed C-terminal diglycine required for SUMO-2 modification and therefore may be suitable for in vitro SUMOylation. Captured proteins can be eluted readily for identification, analysis or subsequent biochemical reaction.




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.5 mL UW0100-0500 89165-096 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/peptide name SUMO2
Protein/peptide type Recombinant
Species Human
Conjugation Agarose
Protein type Recombinant
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping