About this item
ECL Direct Nucleic Acid Labeling and Detection Systems are based on the direct labeling of DNA or RNA probes with horseradish peroxidase (HRP) in a simple 20 minute chemical reaction. The resulting probe can be used without purification. Detection is achieved by generation of light via the HRP-catalyzed breakdown of luminol.
- Direct probe labelling in a 10 minute reaction, 1 hour from hybridization to detection with ECL Direct and Hybond™ N+.
- Eliminates handling, waste and regulatory issues associated with use of radioactivity.
- No need to strip blots before reprobing.
- Contains a novel rate enhancer, giving higher sensitivity with very low backgrounds on nylon membranes.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Related Information
sds_RPN3000.pdf sds_RPN3000A.pdf sds_RPN3000B.pdf sds_RPN3000C.pdf sds_RPN3000D.pdf sds_RPN3000E.pdf sds_RPN3000F.pdf sds_RPN3000G.pdf sds_RPN3000H.pdf sds_RPN3001.pdf sds_RPN3001A.pdf sds_RPN3001B.pdf sds_RPN3001C.pdf sds_RPN3001D.pdf sds_RPN3001E.pdf sds_RPN3001F.pdf sds_RPN3001G.pdf sds_RPN3001H.pdf
SDS
CyDye Licensing Statements GE Healthcare Life Sciences Cytiva CyDye Licensing Statements Cytiva General Licensing Statements Licensing Statements GE Healthcare Life Sciences
Additional Product Details
25006549_HAND BOOK.pdf 25006549_HAND BOOK.pdf 25006549_INSTRUCTIONS.1.pdf 25006549_INSTRUCTIONS.1.pdf 25006549_INSTRUCTIONS.2.pdf 25006549_INSTRUCTIONS.2.pdf 25006549_INSTRUCTIONS.3.pdf 25006549_INSTRUCTIONS.3.pdf 25006549_INSTRUCTIONS.4.pdf 25006549_INSTRUCTIONS.4.pdf 25006549_INSTRUCTIONS.pdf 25006549_INSTRUCTIONS.pdf 25006549_USER MANUAL.pdf 25006549_USER MANUAL.pdf 25006550_HAND BOOK.pdf 25006550_HAND BOOK.pdf 25006550_INSTRUCTIONS.pdf 25006550_INSTRUCTIONS.pdf AM-litdocnaldcat_20110830221542.pdf AR-litdocS5465389_20130721222904.pdf AR-litdocS5465389_2_20130721222843.pdf AT-litdocRPN3000_Rev_E_2006_web_20110830232546.pdf AT-litdoctt125_20110830190514.pdf AT-litdoctt134_20110830172751.pdf