Ficoll PM 70, Cytiva
Supplier: Cytiva
Certificates
About this item
Ficoll® PM70 is a high molecular weight sucrose polymer formed by copolymerisation of sucrose with epichlorohydrin. The molecules are highly branched and the high content of hydroxyl groups leads to very good solubility in aqueous media.
- Displays minimal toxicity
- Favourable viscosity and osmotic properties
The molecules are highly branched and the high content of hydroxyl groups leads to very good solubility in aqueous media.
Specifications
- Catalog No:
- CA89128-846
- CA89128-848
- Endotoxin activity:
- 0.12 EU/mg (Max.)
- 0.12 EU/mg (Max.)
- Molecular weight:
- ~ 6 × 104 [Mr] Min.; ~ 8 × 104 [Mr] Max.
- ~ 6 × 104 [Mr] Min.; ~ 8 × 104 [Mr] Max.
- Radius:
- ~ 5.1 nm (Stokes Radius)
- ~ 5.1 nm (Stokes Radius)
- Rotation:
- 50°-65° (Specific Rotation [alpha]20)
- 50°-65° (Specific Rotation [alpha]20)
- Viscosity:
- ~ 10 ml/g (Intrinsic Viscosity)
- ~ 10 ml/g (Intrinsic Viscosity)
- Shelf life:
- 5 years (Estimated Shelf Life from Manufacture Date)
- 5 years (Estimated Shelf Life from Manufacture Date)
- Storage conditions:
- 4–30°C
- 4–30°C
Specifications
Related Information
CyDye Licensing Statements GE Healthcare Life Sciences Cytiva CyDye Licensing Statements Cytiva General Licensing Statements Licensing Statements GE Healthcare Life Sciences
Additional Product Details
89128-848_10297022
Conformity declaration
Customers Who Bought This Also Bought