Product 30621-392 is similar to Competitor's number 80080-597, but manufacturer and specifications may vary.
Product 89203-512 is similar to Competitor's number 80080-597, but manufacturer and specifications may vary.
Product 89203-512 is similar to Competitor's number 80080-597, but manufacturer and specifications may vary.
Specifications
- Dimensions:140W×140L×54H mm (5 1/2×5 1/2×2 1/8")
- Description:Black, 100-Place, Hinged Polypropylene Cryobox
Specifications
About this item
Ideal for light-sensitive specimens, this all-black storage box is suitable for use with 0.5, 1.5, and 2.0 mL microtubes, as well as most other brands of cryogenic vials.
- Polypropylene fabrication
- Protects light-sensitive specimens
- Lid features an alphanumeric grid
Polypropylene box can accommodate several sizes of cryogenic vials, or tubes of similar diameters. The lid has an alphanumeric grid reference to aid with sample identification. Other matte surfaces are suitable for marking or labeling. The box base and lid have impressed rims for stacking.