Isobutyl chloroformate for synthesis, Sigma-Aldrich®
Catalog #: CA8.02358.0100
Supplier: MilliporeSigma
Price
103,17 $
Log in for availability
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass Bottle
- Size:100 mL
- Size:100 mL
Specifications
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Molar Mass:136.58 g/mol
- Boiling Point:127 - 129 °C (1013 hPa)
- Flash Point:30 °C
- Refractive Index:1.407 (20 °C, 589 nm)
- Density:1.05 g/cm3 (20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
- Sigma-Aldrich®, Isobutyl chloroformate for synthesis:
Specification Test Results
About this item
Synonyms: Chloroformic acid isobutyl ester
Boiling Pt:128.7 °C (1013 hPa)
Melting Pt:–80 °C
Density:1.05 g/cm³ (20 °C)
ADR:6.1 (3, 8),II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:M-Clarity™ quality level = MQ200
Flash Pt:30 °C
:Boiling Pt:
:Boiling Pt: 128.7 °C (1013 hPa)|Boiling Pt: 128.7 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –80 °C
:Flash Pt:
:Flash Pt: 30 °C|Flash Pt: 30 °C
:M.W. 136.58 g/mol
Merck Index:13,05156
:Isobutyl chloroformate
Formula:C₅H₉ClO₂
MW:136.58 g/mol
MDL Number:MFCD00000642
EINECS:208-840-1
:-80|–80 °C
:128.7
CAS Number:543-27-1
UN:2742
:6.1 (3, 8)
:II
:30
:GHS02
:GHS06
:GHS05
:MQ200
:Melting Pt: -80 °C|Melting Pt: –80 °C
Customers Who Bought This Also Bought