EDTA 0.714 M, cartridge for digital titrator

Supplier: Hach
Warning

Synonyms: Ethylenediaminetetraacetic acid tetrasodium saltTetrasodium ethylenediaminetetraacetateEthylenedinitrilotetra-acetic acid tetrasodium saltEDTA tetrasodiumEdetic acid tetrasodium saltKestranal® 4S

1495901 14959-01
CA14959-01EA 61.21 CAD
CA14959-01 97026-528
EDTA 0.714 M, cartridge for digital titrator
EDTA tetrasodium salt

EDTA Cartridge for Digital Titrator for the determination of Total Hardness.


  • Accurate results - when and where you need them
  • Fast and Flexible
  • Versatile
  • Economical


0.714 M. Digits x 1.785 = mg/L CaCO3 in 50 mL, Digits x 0.1 = Gdh.

Formula: C₁₀H₁₂N₂Na₄O₈
MW: 380.17 g/mol
MDL Number: MFCD00150027
CAS Number: 64-02-8
https://ca.vwr.com/stibo/web/std.lang.all/59/84/2355984.jpg

Order Now

Specification Test Results

Method 8204
Method Name EDTA (CalVer® 2)
Number of tests 40 - 100
Parameter Gesamt-Härte, sequentiell
Platform Digital Titrator
Range 10 - 200 G.d.h.
Titrant 0.714 M EDTA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR