Model CY-3 Cyanuric Acid Test Kit, Hach
Supplier: Hach
185102 185102
CA89500-664EA 287.2 CAD
CA89500-664 89500-664
Model CY-3 Cyanuric Acid Test Kit, Hach
Environmental Test Kits
A stabilizing agent for chlorine, cyanuric acid is used in swimming pool treatment
Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field
Cyanuric Acid Test Kit measures the turbidity formed in proportion to Cyanuric Acid concentration, after the reagent is added. Contains 100 tests.
Ordering information: One reagent, clippers, graduated cylinder, sample cell with cap, dipstick, instructions and carrying case included