Lambda™ Plus Single-Channel Pipettor, Corning®
Supplier: Corning Life Sciences
4070 4073 4074 4075 4071 4072
89133-478CS 551.85 CAD
89133-478 89133-484 89133-486 89133-488 89133-480 89133-482
Lambda™ Plus Single-Channel Pipettor, Corning®
Pipets Multi Channel Pipettors
Lambda™ Plus Pipettors have been engineered to provide the highest levels of accuracy, reproducibility, and comfort.
  • Feature smooth plunger movement and extremely low pipetting forces
  • A key is included with each pipettor for easy in-house calibration
Volume can be set by turning either the thumb wheel or the plunger button, and a locking mechanism prevents accidental change. The Lambda™ Plus Pipettors have been designed to maintain their calibration, even after repeated use and autoclaving.
Certifications: ISO 8655 standards.
Ordering information: Three-year manufacturer warranty.