Slide Staining Products, Globe Scientific
Supplier: Globe Scientific
513220 513222 513224
89510-842EA 162.79 CAD
89510-842 89510-844 89510-840
Slide Staining Products, Globe Scientific
Staining Boxes
These slide staining products from Globe Scientific offer three great products to simplify the staining process
The staining dish (89510-842) is designed for use with our staining rack and features a lid that holds up to 28 slides for upright drying.
The storage box (89510-844) is produced from polystyrene (PS) and holds up to 4 staining racks (100 slides). The inside lid of the box contains a detailed slide index sheet to keep slides organized.
The staining rack (89510-840) is produced from polyoxymethylene (POM) and is designed for the simultaneous staining of up to 25 microscope slides.