2,4-Dichloro-5-methylpyridine 96%

Supplier: Thermo Scientific

Danger

Synonyms: 2,4-Dichloro-5-picoline

H61634-MD H61634-MD H61634-03 H61634-03
CAAAH61634-MDEA 113.4 CAD
CAAAH61634-MD AAH61634-MD CAAAH61634-03 AAH61634-03
2,4-Dichloro-5-methylpyridine 96%
2,4-Dichloro-5-methylpyridine
Formula: C₆H₅Cl₂N
Storage Temperature: Ambient
MDL Number: MFCD08704813
CAS Number: 56961-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR