Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium-D(+)-glucuronate monohydrate for synthesis

Supplier: MilliporeSigma

Synonyms: D(+)-Glucuronic acid, sodium salt, monohydrate

8.14723.0010 8.14723.0010
CA8.14723.0010 EM8.14723.0010
Sodium-D(+)-glucuronate monohydrate for synthesis


Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point 137 - 141 °C
Molar Mass 234.14 g/mol


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty