Eppendorf® MTP 384-Well Microplates
Supplier: Eppendorf
951040589 951040421 951040341 951040601 951040367 951040503 951040481 951040383 951040464
75874-682CS 4899.6 CAD
89107-722 75874-682 89107-710 89107-712 75874-680 89107-714 89107-716 89107-708 89107-718
Eppendorf® MTP 384-Well Microplates
Microplates HTS Microplates
These Eppendorf Polypropylene Microplates are ideal for storing substances or for applications requiring high resistance to solvents, such as DMSO.
The easy to read OptiTrack® alphanumeric labeling on each plate makes identifying individual wells incredibly easy, accelerates the filling and significantly reduces the risk of error.
The high level of transparency, unique among polypropylene microplates, ensures that you always find your samples at a glance.
For applications where maximum sample recovery is particularly important, the Microplates 384 are available in Protein and DNA LoBind quality.