MOPS/EDTA Buffer, Molecular Biology Grade, MilliporeSigma
Supplier: MilliporeSigma
475916-100ML 475916-100ML
CA80058-448EA 470.43 CAD
80058-448 CA80058-448
MOPS/EDTA Buffer, Molecular Biology Grade, MilliporeSigma
Buffers Electrophoresis Buffers Running Buffers
MOPS/EDTA Buffer is prepared with DEPC for RNA applications
10X liquid concentrate contains 200 mM MOPS, 50 mM sodium acetate, 10 mM EDTA.