Acrodisc® Syringe Filters with Supor® Membrane, Nonsterile, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Acrodisc®
4661 4659 4668 4506 AP-4426 4662 4508 4661 4655 4504 4692 4653 4692 4506 AP-4799 4668 4508 AP-4425 4653 4504 4660 4662 4655 4509 AP-4425 4660 AP-4426 4659 AP-4799
28143-305EA 3481.99 CAD
28143-301 28143-303 28143-305 CA34181-078 CA97035-174 CA97035-176 34181-088 34181-086 CA28139-710 97027-954 CA28143-372 28143-044 CA28143-374 28143-046 CA28143-046 CA28143-048 CA28143-301 CA28143-303 CA34181-086 CA34181-088 28139-710 34181-078 CA97027-954 97035-178 97035-176 97035-174 28143-372 28143-374 CA28143-044
Acrodisc® Syringe Filters with Supor® Membrane, Nonsterile, Cytiva (Formerly Pall Lab)
Filters Syringe Filters
These syringe filters are ideal for biological and general research.
  • Large pore sizes for high throughput and fast flow rates
  • Low protein binding to minimize sample loss
  • Built in pre-filter for difficult to filter liquids
Supor® (polyethersulfone) membrane provides high throughput and fast flow rate. The filters of 0.2 and 0.45 μm pore sizes provide sterilization of small volumes of buffers, culture media, and additives. Low protein binding to minimize sample loss.
These Supor syringe filters are offered as recommended replacements for the discontinued HT Tuffyrn syringe filters.
Certifications: Acrodisc PSF syringe filters are automation certified to assure smooth operation and worry-free performance 24 hours a day in automated workstations.