Square Cover Glasses No. 2, Corning®
Supplier: CORNING CA
2855-18 2855-25 2855-22
89239-702CS 196.76 CAD
89239-702 89239-706 89239-704
Square Cover Glasses No. 2, Corning®
Cover Glasses
These square cover glasses are made from borosilicate glass
They are nonsterile.