Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Transpore™ Tape, 3M™

Supplier: 3M Healthcare
70-2007-3958-2 1527-2 1527-1/2 1527-0 70200739590 MMM 1527-1
Transpore™ sergical tape is a latex-free, hypoallergenic, transparent and perforated plastic tape that offers strong adhesion

The easy, straight, bi-directional tear allows clinicians to tailor tape to each use. Tape is also water-resistant.

Transpore tape is easy to tear into very thin strips, and is easy to use even with gloves

Ordering information: Tape is sold on a standard roll.



 

ORDER

Expand All / Collapse All
Dimensions Supplier No. VWR Catalog Number Unit Price Quantity
1.25 cm x 9.14 m 1527-1/2 CA56222-104 Box of 24 Retrieving Restricted
Case of 240 Retrieving Restricted
2.5 cm x 9.14 m 70-2007-3958-2 CA11310-668 Box of 12 Retrieving Restricted
Case of 120 Retrieving Restricted
5 cm x 9.14 m 70200739590 CA56222-106 Box of 6 Retrieving Restricted
Case of 60 Retrieving Restricted
restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

MDL No. MFCD00133992
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping