Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acclaim C30 LC Columns, Thermo Scientific

Supplier: Thermo Scientific
075721 075722 075723 075724 075725 075726 078662 078661 078664 078663 078666 078665 075720 078660 075718 075719
For separations of hydrophobic, structurally related isomers, Thermo Scientific™ Acclaim C30 reversed-phase columns provide superior results

The covalent modification of C30 alkyl silane on ultrapure, spherical, porous silica gel gives these columns high shape selectivity. They are fully compatible with various aqueous buffers, useful for a broad application range of analytes, and offer more flexibility in method development.

Ordering information: Guard Cartridges require use of the corresponding guard holder. See Acclaim Guard Kit Holder and Coupler V-2 (VWR Part No. 10046-750).



 

ORDER

Expand All / Collapse All
Column Format I.D. x L Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
Analytical 3 x 50 mm 3 µm 200 Å 078663 10044-854 Each Retrieving
Analytical 2.1 x 100 mm 3 µm 200 Å 078665 10044-858 Each Retrieving
Analytical 2.1 x 50 mm 3 µm 200 Å 078666 10044-860 Each Retrieving
Analytical 3 x 250 mm 3 µm 200 Å 075726 10044-788 Each Retrieving
Analytical 4.6 x 150 mm 5 µm 200 Å 075719 10044-774 Each Retrieving
Analytical 2.1 x 250 mm 3 µm 200 Å 078664 10044-856 Each Retrieving
Analytical 3 x 150 mm 3 µm 200 Å 075724 10044-784 Each Retrieving
Analytical 3 x 100 mm 3 µm 200 Å 078662 10044-852 Each Retrieving
Analytical 4.6 x 250 mm 5 µm 200 Å 075718 10044-772 Each Retrieving
Analytical 4.6 x 100 mm 5 µm 200 Å 078660 10044-848 Each Retrieving
Analytical 2.1 x 150 mm 3 µm 200 Å 075725 10044-786 Each Retrieving
Analytical 4.6 x 150 mm 3 µm 200 Å 075723 10044-782 Each Retrieving
Analytical 4.6 x 50 mm 3 µm 200 Å 078661 10044-850 Each Retrieving
Guard Cartridge 2.1 x 10 mm 5 µm 200 Å 075722 10044-780 Each Retrieving
Guard Cartridge 4.6 x 10 mm 5 µm 200 Å 075720 10044-776 Each Retrieving
Guard Cartridge 3 x 10 mm 5 µm 200 Å 075721 10044-778 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description 10044-854 10044-858 10044-860 10044-788 10044-774 10044-780 10044-856 10044-776 10044-784 10044-778 10044-852 10044-772 10044-848 10044-786 10044-782 10044-850
Carbon load 13%
pH range 2 to 8
Surface area 200 m²/g
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty