Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biobasic 18 HPLC Columns, Thermo Scientific

Supplier: Thermo Scientific
73205-100367 72105-101030 72105-052130 72105-150565 72105-150365 72105-150165 72105-159570 72105-100265 72105-102168 72105-100266 72105-159370 72105-100267 72105-104668 72105-030315 72105-030515 72105-154630 72105-051030 72105-253030 72105-259570 72105-259370 72105-011001 72105-059270 72105-059070 72105-152130 72105-154030 72105-103030 72105-309070 72105-154668 72105-039022 72105-254630 72105-014003 72105-012101 72105-014001 72105-252130 72105-012103 72105-152168 72105-150265 72105-159270 72105-012106 72105-100365 72105-150065 72105-151030 72105-100565 72105-050365 72105-050565 72105-259070 72105-102130 72105-030215 72105-019023 72105-059570 72105-251030 72105-252168 72105-013001 72105-059370 72105-104630 72105-254668
For outstanding separation of small-to-medium size peptides, choose Thermo Scientific™ BioBasic™ 18 HPLC Columns

BioBasic reversed-phase columns provide superior chromatography for life science applications because the extra dense bonding chemistry used for these packings produces a highly stable, reproducible surface for reliable results.

Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1mm ID require the 1mm UNIGUARD Holder (10046-746), 2.1/3mm ID require the 2/3mm UNIGUARD Holder (10046-748) and 4/4.6mm ID require the 4/4.6mm UNIGUARD Holder (10046-744).



 

ORDER

Expand All / Collapse All
Description Column Format I.D. x L Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
Columns Analytical 2.1 x 50 mm 5 µm 300 Å 72105-052130 10043-478 Each Retrieving
Columns Analytical 1 x 250 mm 5 µm 300 Å 72105-251030 10046-482 Each Retrieving
Columns Analytical 4.6 x 250 mm 5 µm 300 Å 72105-254630 10046-490 Each Retrieving
Columns Analytical 1 x 50 mm 5 µm 300 Å 72105-051030 10043-476 Each Retrieving
Columns Analytical 2.1 x 100 mm 5 µm 300 Å 72105-102130 10043-504 Each Retrieving
Columns Capillary 0.32 x 150 mm 5 µm 300 Å 72105-150365 10046-460 Each Retrieving
Columns Analytical 1 x 100 mm 5 µm 300 Å 72105-101030 10043-502 Each Retrieving
Columns Capillary 0.18 x 100 mm 5 µm 300 Å 72105-100265 10043-492 Each Retrieving
Columns Capillary 0.5 x 100 mm 5 µm 300 Å 72105-100565 10043-500 Each Retrieving
Columns Capillary 0.1 x 150 mm 5 µm 300 Å 72105-150165 10046-456 Each Retrieving
Columns Analytical 1 x 150 mm 5 µm 300 Å 72105-151030 10046-464 Each Retrieving
Columns Analytical 4.6 x 150 mm 5 µm 300 Å 72105-154630 10046-472 Each Retrieving
Columns Capillary 0.5 x 150 mm 5 µm 300 Å 72105-150565 10046-462 Each Retrieving
Columns Capillary 0.18 x 150 mm 5 µm 300 Å 72105-150265 10046-458 Each Retrieving
Columns Analytical 2.1 x 250 mm 5 µm 300 Å 72105-252130 10046-484 Each Retrieving
Columns Analytical 2.1 x 150 mm 5 µm 300 Å 72105-152130 10046-466 Each Retrieving
Guard Cartridges Guard Cartridge 4 x 10 mm 5 µm 300 Å 72105-014001 10043-456 Each Retrieving
Guard Cartridges Guard Cartridge 1 x 10 mm 5 µm 300 Å 72105-011001 10043-446 Each Retrieving
Guard Cartridges Guard Cartridge 0.5 x 30 mm 5 µm 300 Å 72105-030515 10043-468 Each Retrieving
Guard Cartridges Guard Cartridge 2.1 x 10 mm 5 µm 300 Å 72105-012101 10043-448 Each Retrieving
Guard Cartridges Guard Cartridge 10 x 10 mm 5 µm 300 Å 72105-019023 10043-460 Each Retrieving
PEEK Bio-Inert Column Analytical 2.1 x 100 mm 5 µm 300 Å 72105-102168 10043-506 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description 10043-474 10043-478 10043-456 10043-470 10046-482 10046-490 10043-476 10043-496 10043-504 10043-512 10046-460 10043-488 10046-468 10046-476 10046-492 10043-520 10043-446 10043-454 10043-458 10043-468 10043-494 10043-502 10043-486 10043-484 10043-492 10043-500 10046-456 10046-464 10046-472 10046-488 10046-480 10043-516 10043-450 10043-472 10043-498 10043-464 10043-506 10046-454 10046-462 10043-490 10046-470 10046-494 10043-750 10046-478 10046-486 10043-466 10043-448 10043-460 10043-518 10046-474 10043-452 10043-508 10043-510 10046-458 10046-484 10046-466
Carbon load 9%
pH range 2 to 9
Surface area 100 m²/g
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty