Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biobasic AX HPLC Columns, Thermo Scientific

Supplier: Thermo Scientific
73105-102130 73105-154630 73105-022151 73105-022130 73105-152130 73105-051030 73105-259070 73105-053030 73105-011001 73105-013001 73105-054630 73105-104630 73105-151030 73105-103030 73105-101030 73105-153030 73105-254030 73105-150565 73105-052130 73105-022106 73105-014006 73105-012101 73105-014001 73105-252130 73105-254630 73105-012106
For the separation of proteins, peptides, oligonucleotides and other anionic species, choose Thermo Scientific™ BioBasic AX columns

These high performance columns have a weak anion exchange phase based on polyethyleneimine chemistry. They are ideal for the analysis of proteins, peptides, and oligonucleotides and can also be used for HILIC applications.

Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1mm ID require the 1mm UNIGUARD Holder (10046-746), 2.1/3mm ID require the 2/3mm UNIGUARD Holder (10046-748) and 4/4.6mm ID require the 4/4.6mm UNIGUARD Holder (10046-744).



 

ORDER

Expand All / Collapse All
Description Column Format I.D. x L Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
Cartridge Guard Cartridge 1 x 10 mm 5 µm 300 Å 73105-011001 10043-668 Each Retrieving
Cartridge Guard Cartridge 3 x 10 mm 5 µm 300 Å 73105-013001 10043-674 Each Retrieving
Cartridge Guard Cartridge 2.1 x 10 mm 5 µm 300 Å 73105-012101 10043-670 Each Retrieving
Cartridge Guard Cartridge 4 x 10 mm 5 µm 300 Å 73105-014001 10043-676 Each Retrieving
Column Analytical 2.1 x 50 mm 5 µm 300 Å 73105-052130 10043-688 Each Retrieving
Column Analytical 2.1 x 100 mm 5 µm 300 Å 73105-102130 10043-696 Each Retrieving
Column Analytical 1 x 50 mm 5 µm 300 Å 73105-051030 10043-686 Each Retrieving
Column Analytical 3 x 50 mm 5 µm 300 Å 73105-053030 10043-690 Each Retrieving
Column Analytical 4.6 x 50 mm 5 µm 300 Å 73105-054630 10043-692 Each Retrieving
Column Analytical 4.6 x 100 mm 5 µm 300 Å 73105-104630 10043-700 Each Retrieving
Column Analytical 4.6 x 250 mm 5 µm 300 Å 73105-254630 10043-716 Each Retrieving
Column Analytical 1 x 150 mm 5 µm 300 Å 73105-151030 10043-704 Each Retrieving
Column Capillary 0.5 x 150 mm 5 µm 300 Å 73105-150565 10043-702 Each Retrieving
Column Analytical 2.1 x 150 mm 5 µm 300 Å 73105-152130 10043-706 Each Retrieving
Column Analytical 2.1 x 20 mm 5 µm 300 Å 73105-022130 10043-682 Each Retrieving
Column Analytical 3 x 150 mm 5 µm 300 Å 73105-153030 10043-708 Each Retrieving
Column Preparative 10 x 250 mm 5 µm 300 Å 73105-259070 10043-718 Each Retrieving
Column Analytical 1 x 100 mm 5 µm 300 Å 73105-101030 10043-694 Each Retrieving
Column Analytical 4.6 x 150 mm 5 µm 300 Å 73105-154630 10043-710 Each Retrieving
HTS Column Analytical 2.1 x 20 mm 5 µm 300 Å 73105-022151 10043-684 Each Retrieving
Javelin Guard Guard Column 2.1 x 10 mm 5 µm 300 Å 73105-012106 10043-672 Each Retrieving
Javelin Guard Guard Column 2.1 x 20 mm 5 µm 300 Å 73105-022106 10043-680 Each Retrieving
Javelin Guard Guard Column 4 x 10 mm 5 µm 300 Å 73105-014006 10043-678 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description 10043-672 10043-680 10043-688 10043-696 10043-678 10043-686 10043-690 10043-698 10043-714 10043-668 10043-692 10043-700 10043-716 10043-704 10043-712 10043-702 10043-706 10043-674 10043-682 10043-708 10043-670 10043-684 10043-676 10043-718 10043-694 10043-710
Carbon load 3%
pH range 2 to 8
Surface area 100 m²/g
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty