Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Accucore Urea-HILIC Columns, Thermo Scientific

Supplier: Thermo Scientific
Accucore™
27726-252130 27726-052130 27726-153030 27726-152130 27726-054630 27726-012105 27726-102130 27726-154630 27726-053030 27726-103030 27726-104630
Separate polar analytes with improved speed and resolution using Thermo Scientific™ Accucore Urea-HILIC columns

Rugged 2.6µm solid core particles enable fast, high-resolution separations at low backpressures. The Urea-HILIC bonded hydrophilic stationary phase offers unique selectivity and has lower ion exchange activity compared to other HILIC phases. Robust bonding technology and tightly packed beds ensure excellent reproducibility and long column lifetimes.

Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1mm ID require the 1mm UNIGUARD Holder (10046-746), 2.1/3mm ID require the 2/3mm UNIGUARD Holder (10046-748) and 4/4.6mm ID require the 4/4.6mm UNIGUARD Holder (10046-744)




 

ORDER

Expand All / Collapse All
Column Format I.D. x L Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
Analytical 2.1 x 250 mm 2.6 µm 80 Å 27726-252130 10039-058 Each Retrieving
Analytical 3 x 100 mm 2.6 µm 80 Å 27726-103030 10039-068 Each Retrieving
Analytical 2.1 x 150 mm 2.6 µm 80 Å 27726-152130 10039-064 Each Retrieving
Analytical 2.1 x 50 mm 2.6 µm 80 Å 27726-052130 10039-076 Each Retrieving
Analytical 3 x 150 mm 2.6 µm 80 Å 27726-153030 10039-062 Each Retrieving
Analytical 4.6 x 100 mm 2.6 µm 80 Å 27726-104630 10039-066 Each Retrieving
Analytical 2.1 x 100 mm 2.6 µm 80 Å 27726-102130 10039-070 Each Retrieving
Analytical 3 x 50 mm 2.6 µm 80 Å 27726-053030 10039-074 Each Retrieving
Analytical 4.6 x 150 mm 2.6 µm 80 Å 27726-154630 10039-060 Each Retrieving
Guard cartridge 2.1 x 10 mm 2.6 µm 80 Å 27726-012105 10039-078 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description 10039-058 10039-078 10039-068 10039-064 10039-076 10039-062 10039-066 10039-070 10039-074 10039-060
pH range 2 to 8
Surface area 130 m²/g

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty