Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acclaim Explosives E2 LC Columns, Thermo Scientific

Supplier: Thermo Scientific
076227 070083 070081 071989 070082 064309 069703 076225 076226
Use Thermo Scientific™ Acclaim Explosives E1 and E2 columns to achieve baseline resolution of all 14 explosives listed in EPA SW-846 Method 8330, Nitroaromatics and Nitramines by HPLC

Their unique column chemistries deliver superior resolution with complementary selectivities. Use the E1 column as a direct replacement for ODS columns for primary analysis. Then, use the E2 column as either a primary or a confirmatory column. Easily change the separation conditions for the E2 column to accommodate a wider application range, including the analysis of explosives beyond U.S. EPA Method 8330.

Ordering information: Guard Cartridges require use of the corresponding guard holder. See Acclaim Guard Kit Holder and Coupler V-2 (VWR Part No. 10046-750).



 

ORDER

Expand All / Collapse All
Column Format I.D. x L Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
Analytical 3 x 150 mm 3 µm 120 Å 070082 10044-668 Each Retrieving
Analytical 2.1 x 150 mm 2.2 µm 120 Å 076226 10044-812 Each Retrieving
Analytical 3 x 250 mm 3 µm 120 Å 070081 10044-666 Each Retrieving
Analytical 2.1 x 100 mm 2.2 µm 120 Å 076225 10044-810 Each Retrieving
Analytical 4.6 x 250 mm 5 µm 120 Å 064309 10044-562 Each Retrieving
Analytical 3 x 100 mm 2.2 µm 120 Å 076227 10044-814 Each Retrieving
Analytical 2.1 x 150 mm 3 µm 120 Å 070083 10044-670 Each Retrieving
Guard Cartridge 4.6 x 10 mm 5 µm 120 Å 069703 10044-650 Each Retrieving
Guard Cartridge 3 x 10 mm 5 µm 120 Å 071989 10040-550 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description 10044-668 10044-812 10044-666 10044-810 10044-650 10044-562 10044-814 10040-550 10044-670
pH range 3 to 7
Surface area 300 m²/g
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping