Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acclaim Carbamate LC Columns, Thermo Scientific

Supplier: Thermo Scientific
072929 072928 072927 072926 072925 072924 075596 072930 075597
Use with Thermo Scientific™ Acclaim Carbamate LC columns to achieve baseline separation of carbamate pesticides (N-methylcarbamate and N-methylcarbamoyloxime pesticides) specified in US EPA Method 531.2

These high-efficiency columns are compatible with both binary (methanol/water) and ternary (acetonitrile/methanol/water) mobile phase gradients for greater flexibility. Use these extremely low bleed columns with either LC/post-column derivatization/fluorescence or LC/MS detection for the ultimate sensitivity.

Ordering information: Guard Cartridges require use of the corresponding guard holder. See Acclaim Guard Kit Holder and Coupler V-2 (VWR Part No. 10046-750).




 

ORDER

Expand All / Collapse All
Column Format I.D. x L Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
Analytical 2.1 x 150 mm 2.2 µm 120 Å 075596 10044-756 Each Retrieving
Analytical 2.1 x 100 mm 2.2 µm 120 Å 075597 10044-758 Each Retrieving
Analytical 3 x 150 mm 3 µm 120 Å 072926 10040-586 Each Retrieving
Analytical 4.6 x 150 mm 3 µm 120 Å 072925 10040-584 Each Retrieving
Analytical 2.1 x 150 mm 3 µm 120 Å 072927 10040-588 Each Retrieving
Analytical 4.6 x 250 mm 5 µm 120 Å 072924 10040-582 Each Retrieving
Guard cartridge 4.6 x 10 mm 3 µm 120 Å 072928 10040-590 Each Retrieving
Guard cartridge 2.1 x 10 mm 3 µm 120 Å 072930 10040-594 Each Retrieving
Guard cartridge 3 x 10 mm 3 µm 120 Å 072929 10040-592 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description 10044-756 10044-758 10040-590 10040-594 10040-586 10040-592 10040-584 10040-588 10040-582
pH range 2.5 to 8
Surface area 300 m²/g

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty