Anti-FASLG Rabbit Polyclonal Antibody

48-479
10078-130EA 780.2 CAD
10078-130
Anti-FASLG Rabbit Polyclonal Antibody
Apoptosis antigen ligand 1 is the ligand for FAS. Both are transmembrane proteins. Interaction of FAS with its ligand is critical in triggering apoptosis of some types of cells such as lymphocytes. Defects in TNFSF6 may be related to some cases of systemic lupus erythematosus (SLE).

FASLG antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

PBS, pH 7.4, 0.2% BSA, 0.05% sodium azide.

Aliquot and store at 4ºC. Avoid freezing.

Type:
Antigen: FASLG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR