Anti-GZMB Rabbit Polyclonal Antibody

48-488
10078-136EA 623.55 CAD
10078-136
Anti-GZMB Rabbit Polyclonal Antibody
GZMB antibody can be used in Western Blot, and immunohistochemistry starting at 1:100.

PBS, pH 7.4, 0.2% BSA, 0.05% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GZMB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR