Anti-ATF2 Rabbit Polyclonal Antibody

48-503
10078-156EA 1011.8 CAD
10078-156
Anti-ATF2 Rabbit Polyclonal Antibody
Antibodies
  • ATF2 antibody can be used in immunohistochemistry starting at 20 μg/mL, and flow cytometry.

PBS, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ATF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR