Anti-BCL2 Rabbit Polyclonal Antibody

48-505
10078-160EA 811.46 CAD
10078-160
Anti-BCL2 Rabbit Polyclonal Antibody
Bcl-2 inhibits apoptosis. Translocations that lead to overexpression of this gene are associated with cancer (lymphoma in the case of translocation to the Ig heavy chain locus).

BCL2 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

PBS, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BCL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR