Anti-POU5F1 Rabbit Polyclonal Antibody

48-522
10078-194EA 664.85 CAD
10078-194
Anti-POU5F1 Rabbit Polyclonal Antibody
POU5F1 is a POU transcription factor expressed by early embryo cells and germ cells. It determines paracrine growth factor signaling from stem cells to the trophectoderm.

POU5F1 antibody can be used in ELISA starting at 1 ?g/mL, Western Blot starting at 1 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

Protein A/G Column

PBS, 2% sucrose.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: POU5F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Goat, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR