Order Entry
ContactUsLinkComponent
Anti-MMP13 Rabbit Polyclonal Antibody
Anti-MMP13 Rabbit Polyclonal Antibody
Catalog #: 10078-214
Supplier:  Avantor
Anti-MMP13 Rabbit Polyclonal Antibody
Catalog #: 10078-214
Supplier:  Avantor
Discontinued
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    matrix metallopeptidase 13 (collagenase 3)
  • Clonality:
    Polyclonal
  • Gene ID:
    P45452
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Antigen symbol:
    MMP13
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Size:
    0.05 mg
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    MMP13
  • Storage buffer:
    PBS, pH 7.4, 0.05% sodium azide.
  • Storage temperature:
    Store MMP13 antibody at 4 °C or –20 °C. As with all antibodies avoid freeze/thaw cycles.
  • Concentration:
    0.2 mg/mL
  • Shipping temperature:
    4 °C
  • Immunogen:
    MMP13 antibody was raised against a peptide from the hinge region of MMP13 (Human).
  • Purification:
    Affinity Chromatography
  • Cat. no.:
    10078-214

Specifications

About this item

MMP13 is a Metallopeptidase M10A type protease. Metalloproteinase 13/Collagenase 3 (MMP13) was originally identified in breast carcinoma and has since been documented in a variety of tumors including chondrosarcomas, basal cell carcinoma, and malignant melanoma. In addition it has been detected in inflammatory diseases such as atherosclerosis, in wound healing, and in destructive joint diseases such OA and RA. MMP13 degrades the native helix of several types of fibrillar collagen, and extracellular matrix proteins in vitro.

MMP13 antibody can be used in ELISA starting at 1:10000, immunohistochemistry starting at 1:200, and immunofluorescence starting at 1:200 - 1:1000.

Affinity Chromatography

PBS, pH 7.4, 0.05% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MMP13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Customers Who Bought This Also Bought