MMP7 is a Metallopeptidase M10A type protease. MMP-7 (Matrilysin) is constitutively produced by exocrine epithelial cells through out the body. It is capable of degrading casein, fibronectin, gelatins of types 1, III, IV, and V, in addition to activating procollagenase. It has been shown to play a key role in invasion by carcinomas.
MMP7 antibody can be used in ELISA starting at 1:10000, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 1:100 - 1:200.
PBS, pH 7.4, 0.1% BSA, 0.05% sodium azide.
Store MMP7 antibody at 2-8 °C for up to 1 year. Avoid repeated freeze-thaw cycles.
Type: Primary
Antigen: MMP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat