Anti-MMP8 Rabbit Polyclonal Antibody

48-569
10078-220EA 620.52 CAD
10078-220
Anti-MMP8 Rabbit Polyclonal Antibody
MMP8 is a Metallopeptidase M10A type protease. MMP8 is a Metallopeptidase M10A type protease. It is synthesized and stored intracellularly in specific granules in neutrophils. MMP8 shows a preference for degrading type I collagen.

MMP8 antibody can be used in ELISA, and immunohistochemistry starting at 20 ?g/mL.

Purified

PBS, pH 7.4, 0.05% sodium azide.

Store MMP8 antibody at 2-8 °C for up to 1 year. Avoid repeated freeze-thaw cycles.

Type:
Antigen: MMP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR