SOCS (Suppressor of cytokine signaling) family proteins form part of a classical negative feedback system that regulates cytokine signal transduction. They are involved in the negative regulation of cytokines that signal through the JAK/STAT pathway. In addition, SOCS2 has been shown to interact with the Insulin-like Growth Factor I Receptor and act as an intracellular mediator of the anti-inflammatory actions of aspirin-induced lipoxins in vivo.
SOCS2 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.
Affinity Chromatography
PBS, pH 7.4, 0.05% sodium azide.
Aliquot and store at 4ºC. Avoid freezing.
Type:
Antigen: SOCS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human