Anti-STEAP1 Rabbit Polyclonal Antibody

48-587
10078-234EA 620.52 CAD
10078-234
Anti-STEAP1 Rabbit Polyclonal Antibody
STEAP1 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 1:200.

1x PBS (pH 7.4) containing 0.1% BSA and 0.05% sodium azide.

Store STEAP1 antibody at 2-8 °C for up to 1 year. Avoid repeated freeze-thaw cycles.

Type:
Antigen: STEAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR