Anti-LEP Rabbit Polyclonal Antibody

Supplier: Prosci
48-684
10078-286EA 955.07 CAD
10078-286
Anti-LEP Rabbit Polyclonal Antibody
Antibodies
Leptin has been implicated in the control of appetite and metabolism.

Leptin antibody can be used in ELISA, and immunohistochemistry starting at 40 ?g/mL.

Antiserum

Contains 0.01% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LEP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR