Anti-CDKN1B Rabbit Polyclonal Antibody

48-690
10078-296EA 811.46 CAD
10078-296
Anti-CDKN1B Rabbit Polyclonal Antibody
CDKN1B antibody can be used in Western Blot, and immunohistochemistry starting at 20 ?g/mL.

PBS, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CDKN1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR