Anti-HSP90AB1 Rabbit Polyclonal Antibody

48-703
10078-318EA 811.46 CAD
10078-318
Anti-HSP90AB1 Rabbit Polyclonal Antibody
HSP90AB1 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 1:200, and flow cytometry starting at 1:200 - 1:400.

PBS, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HSP90AB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR