Anti-TUBG1 Rabbit Polyclonal Antibody

Supplier: Prosci

48-720
10078-346EA 1124.21 CAD
10078-346
Anti-TUBG1 Rabbit Polyclonal Antibody
Antibodies
TUBG1 antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TUBG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR