Anti-HIF1A Rabbit Polyclonal Antibody

48-726
10078-356EA 620.52 CAD
10078-356
Anti-HIF1A Rabbit Polyclonal Antibody
HImmunofluorescence1A antibody can be used in immunohistochemistry starting at 5 ?g/mL.

TBS, pH 7.4, 0.02% sodium azide, 0.5 mg/mL BSA, 50% glycerol.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HIF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bat, Bovine, Chimpanzee, Dog, Gibbon, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Orangutan, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR