Anti-GSTP1 Mouse Monoclonal Antibody [clone: [3F2C2]]

Supplier: Prosci

48-731
10078-362EA 955.07 CAD
10078-362
Anti-GSTP1 Mouse Monoclonal Antibody [clone: [3F2C2]]
Antibodies
GSTP1 antibody can be used in ELISA starting at 1:4000 - 1:16000, Western Blot starting at 1:500 - 1:3000, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.4, 0.03% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GSTP1
Clonality: Monoclonal
Clone: [3F2C2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR