Anti-EAF2 Rabbit Polyclonal Antibody

48-745
10074-368EA 620.52 CAD
10074-368
Anti-EAF2 Rabbit Polyclonal Antibody
EAF2 antibody can be used in ELISA starting at 1:2500 - 1:10000, Western Blot starting at 1:2500 - 1:10000, and immunohistochemistry starting at 5 ?g/mL.

TBS, pH 7.4, 50% glycerol, 0.1% BSA, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: EAF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gibbon, Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR