Anti-LPAR3 Rabbit Polyclonal Antibody

48-746
10074-370EA 620.52 CAD
10074-370
Anti-LPAR3 Rabbit Polyclonal Antibody
EDG7, a Lysophospholipid/Lysosphingolipid Receptor, was identified by RT-PCR from Jurkat T-cells. It is activated by unsaturated lysophosphatidic acid LPA.

EDG7 antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

TBS, pH 7.4, 50% glycerol, 0.1% BSA, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LPAR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR