Anti-NOS1 Rabbit Polyclonal Antibody

Supplier: Prosci

48-752
10074-378EA 1011.8 CAD
10074-378
Anti-NOS1 Rabbit Polyclonal Antibody
Antibodies
NOS1 antibody can be used in ELISA, and immunohistochemistry starting at 5 ?g/mL.

TBS, pH 7.4, 0.5 mg/mL BSA, 50% glycerol, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NOS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bat, Dog, Gorilla, Horse, Human, Monkey, Mouse, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR