Anti-PLA2G5 Rabbit Polyclonal Antibody

48-763
10074-390EA 656.81 CAD
10074-390
Anti-PLA2G5 Rabbit Polyclonal Antibody
PLA2G5 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

TBS, pH 7.4, 0.5% BSA, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PLA2G5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR