Anti-PLA2G2A Mouse Monoclonal Antibody

48-765
10074-394EA 620.52 CAD
10074-394
Anti-PLA2G2A Mouse Monoclonal Antibody
Phospholimmunoprecipitationase A2 antibody can be used in Western Blot starting at 1:1000 - 1:5000, immunohistochemistry starting at 10 ?g/mL, immunocytochemistry starting at 1:100 - 1:500, immunoprecipitation, and flow cytometry.

PBS, pH 7.4, 0.5% mg/mL BSA, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PLA2G2A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR