Anti-PTGIS Rabbit Polyclonal Antibody

48-766
10074-396EA 620.52 CAD
10074-396
Anti-PTGIS Rabbit Polyclonal Antibody
Prostacyclin Synthase antibody can be used in immunohistochemistry starting at 10 ?g/mL.

TBS, pH 7.4, 0.5% BSA, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PTGIS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR