Anti-S100 A8 Mouse Monoclonal Antibody [clone: [CF-145]]

Supplier: Prosci

48-769
10074-402EA 1046.96 CAD
10074-402
Anti-S100 A8 Mouse Monoclonal Antibody [clone: [CF-145]]
Antibodies
Calgranulin A antibody can be used in Western Blot starting at 1 - 2 ?g/mL, and immunohistochemistry starting at 1:50.

PBS, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: S100A8
Clonality: Monoclonal
Clone: [CF-145]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR